Skip to Content
Merck
All Photos(1)

Key Documents

474010

Sigma-Aldrich

(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one

98%

Synonym(s):

(R)-3-Methoxy-1,2-propanediol cyclic carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O4
CAS Number:
Molecular Weight:
132.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
To inquire about this product 474010, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

optical activity

[α]20/D +44°, neat

optical purity

ee: 97% (GLC)

refractive index

n20/D 1.437 (lit.)

bp

105-106 °C/0.9 mmHg (lit.)

density

1.24 g/mL at 25 °C (lit.)

SMILES string

COC[C@@H]1COC(=O)O1

InChI

1S/C5H8O4/c1-7-2-4-3-8-5(6)9-4/h4H,2-3H2,1H3/t4-/m1/s1

InChI key

DNSGQMOSYDHNHO-SCSAIBSYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service