Skip to Content
Merck
All Photos(1)

Key Documents

465356

Sigma-Aldrich

4-(Trifluoromethoxy)anisole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4OCH3
CAS Number:
Molecular Weight:
192.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.432 (lit.)

bp

164 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(OC(F)(F)F)cc1

InChI

1S/C8H7F3O2/c1-12-6-2-4-7(5-3-6)13-8(9,10)11/h2-5H,1H3

InChI key

NOAFZIOGGDPYKK-UHFFFAOYSA-N

General description

4-(Trifluoromethoxy)anisole can undergo metalation preferentially or exclusively at the methoxy-neighboring position.[1][2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

α-Fluorinated ethers, thioethers, and amines: Anomerically biased species.
Leroux F, et al.
Chemical Reviews, 105(3), 827-856 (2005)
Frédéric R Leroux et al.
Beilstein journal of organic chemistry, 4, 13-13 (2008-10-23)
After nitrogen, fluorine is probably the next most favorite hetero-atom for incorporation into small molecules in life science-oriented research. This review focuses on a particular fluorinated substituent, the trifluoromethoxy group, which is finding increased utility as a substituent in bioactives
F Aladar Bencsath et al.
Analytical and bioanalytical chemistry, 407(14), 4079-4090 (2015-03-23)
A headspace solid-phase microextraction gas chromatography-mass spectrometry (SPME GC-MS) method is described, to screen seafood for volatile organic compounds (VOCs) associated with petrochemical taint. VOCs are extracted from the headspace of heated sample homogenates by adsorption onto a SPME fiber

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service