Skip to Content
Merck
All Photos(1)

Key Documents

463884

Sigma-Aldrich

Nickel(II) perchlorate hexahydrate

99.999% trace metals basis

Synonym(s):

Nickel diperchlorate hexahydrate, Nickel perchlorate hexahydrate, Nickelous perchlorate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(ClO4)2 · 6H2O
CAS Number:
Molecular Weight:
365.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

99.999% trace metals basis

form

crystalline

reaction suitability

reagent type: oxidant

mp

140 °C (lit.)

SMILES string

O.O.O.O.O.O.[Ni++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Ni.6H2O/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);;6*1H2/q;;+2;;;;;;/p-2

InChI key

WHGYCGOFTBFDLW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.[1]

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 3 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy C. Higgs et al.
Inorganic chemistry, 37(9), 2263-2272 (2001-10-24)
The mixed functionality pyrazole/phenol ligand (2-hydroxyphenyl)bis(pyrazolyl)methane, L1OH, has been used to prepare a series of linear trimetallic systems with the general structural motif [M(3)(L1O)(4)](2+), where M = Mn(2+), Co(2+), Ni(2+), Cu(2+), and Zn(2+). Each of these complexes has been structurally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service