Skip to Content
Merck
All Photos(1)

Key Documents

461393

Sigma-Aldrich

(S)-β-Methylphenethylamine

99%

Synonym(s):

(S)-2-Phenyl-1-propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)CH2NH2
CAS Number:
Molecular Weight:
135.21
Beilstein:
3195645
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]22/D −35°, c = 1 in ethanol

refractive index

n20/D 1.525 (lit.)

mp

207 °C (lit.)

density

0.945 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

C[C@H](CN)c1ccccc1

InChI

1S/C9H13N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/t8-/m1/s1

InChI key

AXORVIZLPOGIRG-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bacteriorhodopsin is the prototypical "seven-helix" transmembrane protein (with seven α-helical domains), whose study led to advances in understanding G protein-coupled receptors (GPCRs). In Halobacteria, it acts as a light-harvesting protein, producing a proton gradient across the cell wall that is then used to drive biosynthetic processes.

Application

Building block for the enantioselective synthesis of pyrizinostatin, a pyroglutamyl peptidase inhibitor.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kitagawa, M. Tatsuta, K.
Heterocycles, 38, 1747-1747 (1994)
Anita H Lewin et al.
Bioorganic & medicinal chemistry, 16(15), 7415-7423 (2008-07-08)
A cell line in which RD-HGA16 cells were stably transfected with the hTAAR 1 receptor was created and utilized to carry out a systematic evaluation of a series of beta-phenethylamines. Fair agreement was observed with data obtained for aryl and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service