Skip to Content
Merck
All Photos(1)

Key Documents

457760

Sigma-Aldrich

(R)-(−)-N-Benzyl-2-phenylglycinol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHCH(C6H5)CH2OH
CAS Number:
Molecular Weight:
227.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]21/D −80°, c = 0.85 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

bp

87-90 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC[C@H](NCc1ccccc1)c2ccccc2

InChI

1S/C15H17NO/c17-12-15(14-9-5-2-6-10-14)16-11-13-7-3-1-4-8-13/h1-10,15-17H,11-12H2/t15-/m0/s1

InChI key

FTFBWZQHBTVPEO-HNNXBMFYSA-N

Application

Used for the asymmetric synthesis of α-amino phosphonic acids.[1] Starting material for a practical route to optically pure β-substituted amines.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Maury et al.
The Journal of organic chemistry, 61(11), 3687-3693 (1996-05-31)
A simple and general asymmetric synthesis of alpha-amino phosphonic acids is described. The method involves the highly selective addition of trialkyl phosphite onto various chiral oxazolidines. Oxazaphosphorinanes thus obtained with an excellent diastereoselectivity furnish the corresponding (S)-alpha-substituted amino phosphonic acids
Micouin, L. et al.
Tetrahedron Letters, 35, 7223-7223 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service