Skip to Content
Merck
All Photos(1)

Key Documents

449768

Sigma-Aldrich

Chromium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Chromic fluoride, Chromic trifluoride, Chromium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrF3
CAS Number:
Molecular Weight:
108.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

density

3.8 g/mL at 25 °C (lit.)

SMILES string

F[Cr](F)F

InChI

1S/Cr.3FH/h;3*1H/q+3;;;/p-3

InChI key

FTBATIJJKIIOTP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of a mixed-metal fluoride catalyst for synthesis of HFC-134a.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysis Letters, 41, 221-221 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service