Skip to Content
Merck
All Photos(1)

Key Documents

449210

Sigma-Aldrich

Indium(I) chloride

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Chloroindium(I), Indium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InCl
CAS Number:
Molecular Weight:
150.27
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: indium

bp

608 °C (lit.)

mp

225 °C (lit.)

density

4.19 g/mL at 25 °C (lit.)

SMILES string

Cl[In]

InChI

1S/ClH.In/h1H;/q;+1/p-1

InChI key

APHGZSBLRQFRCA-UHFFFAOYSA-M

Application

Used to prepare cage, dinuclear complexes (In-cyclodiphophazanes) which possess interesting metal-metal interactions.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grocholl, L. et al.
Inorganic Chemistry, 37, 2496-2496 (1998)
Chun Kiang Chua et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(7), 1871-1877 (2014-02-04)
Graphene is a promising material capable of driving technological advancement. It is, however, a challenge to obtain pristine graphene in large quantities given the limitation of current synthetic methods. Among the numerous methods available, the chemical approach provides an optimistic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service