Skip to Content
Merck
All Photos(1)

Key Documents

447978

Sigma-Aldrich

Dextrin palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H102O9
CAS Number:
Molecular Weight:
895.38
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

mp

80-130 °C (lit.)

acid number

4‑7 mg KOH/g

saponification value

195‑220 mg KOH/g

density

0.53 g/mL at 25 °C (lit.)

Application

Gelling agent for liquid paraffins, higher alcohols, esters and ketones. Oil phase stabilizer for o/w and w/o systems. Uniformly dispenses dyes.

Features and Benefits

Surfactants have no effect on gel strength. Forms temperature-reversible gels.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service