Skip to Content
Merck
All Photos(1)

Key Documents

443026

Sigma-Aldrich

2-Amino-6-fluorobenzoic acid

98%

Synonym(s):

6-Fluoroanthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(F)CO2H
CAS Number:
Molecular Weight:
155.13
Beilstein:
3540926
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

167-169 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cccc(F)c1C(O)=O

InChI

1S/C7H6FNO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,9H2,(H,10,11)

InChI key

RWSFZKWMVWPDGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of a novel benzamide with potent neuroleptic activity.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Norman et al.
Journal of medicinal chemistry, 39(5), 1172-1188 (1996-03-01)
A series of substituted (4-(4-(1,2-benzisothiazol-3-yl)-1-piperazinyl)butyl)benzamide derivatives was prepared and evaluated as potential atypical antipsychotic agents. The target compounds were readily prepared from their benzoyl chloride, benzoic acid, or isatoic anhydride precursors, and they were evaluated in vitro for their ability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service