Skip to Content
Merck
All Photos(1)

Documents

436135

Sigma-Aldrich

4-Fluoro-2-(trifluoromethyl)benzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)CO2H
CAS Number:
Molecular Weight:
208.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

121-124 °C (lit.)

SMILES string

OC(=O)c1ccc(F)cc1C(F)(F)F

InChI

1S/C8H4F4O2/c9-4-1-2-5(7(13)14)6(3-4)8(10,11)12/h1-3H,(H,13,14)

InChI key

JUHPDXOIGLHXTC-UHFFFAOYSA-N

General description

4-Fluoro-2-(trifluoromethyl)benzoic acid is a substituted benzoic acid derivative. Molecular orbital and empirical methods have been utilized to determine its computed steric and electronic properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott B Hoyt et al.
Bioorganic & medicinal chemistry letters, 17(22), 6172-6177 (2007-09-25)
A series of benzazepinones were synthesized and evaluated as hNa(v)1.7 sodium channel blockers. Several compounds from this series displayed good oral bioavailability and exposure and were efficacious in a rat model of neuropathic pain.
F Y Ghauri et al.
Biochemical pharmacology, 44(10), 1935-1946 (1992-11-17)
An extensive set of computed molecular properties, both steric and electronic, have been calculated using molecular orbital and empirical methods for benzoic acid (1) and a congeneric series of substituted benzoic acids, i.e. 2-, 3- and 4-fluorobenzoic acids (2-4), 2-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service