Skip to Content
Merck
All Photos(2)

Key Documents

43146

Sigma-Aldrich

1,5-Diphenyl-1,3,5-pentanetrione

≥98.0% (T)

Synonym(s):

1,3-Dibenzoylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COCH2COC6H5
CAS Number:
Molecular Weight:
266.29
Beilstein:
2054189
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

mp

107-109 °C

functional group

ketone
phenyl

SMILES string

O=C(CC(=O)c1ccccc1)CC(=O)c2ccccc2

InChI

1S/C17H14O3/c18-15(11-16(19)13-7-3-1-4-8-13)12-17(20)14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

MUNGMRPYTCHBFX-UHFFFAOYSA-N

General description

1,5-Diphenyl-1,3,5-pentanetrione (H2DBA, DBAc) is a symmetrical triketone.1,3,5-triketones are potential tridentate ligands and forms complexes with transition metals, lanthanides and actinides. It acts as ligand and forms binuclear cobalt complex with CoCl2.6H2O.[1] The interaction of two intramolecular hydrogen bonds in 1,5-diphenyl-1,3,5-pentanetrione has been studied.[2] H2DBA can be synthesized by condensation reaction between ethyl benzoate and benzoyl acetone.[3] DBAc arranged in stretched polyethylene (PE) sheets has been investigated by infrared linear dichroism (LD) spectroscopy.[4]

Other Notes

Versatile triketone, used e.g. for the synthesis of heterocyclic compounds; Preparation of complexes with transition metals [5][6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infrared linear dichroism on 1,5-diphenyl-1,3,5-pentanetrione aligned in stretched polyethylene.
Andersen KB.
Acta Chemica Scandinavica, 52, 1171-1176 (1998)
Electronic states of 1, 6, 6aλ4-trithiapentalene and its 2,5-dimethyl and 2,5-diphenylderivatives. Ultraviolet-visible linear dichroism spectroscopy and time-dependent density functional theory calculations.
Spanget-Larsen J.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 58(10), 2245-2255 (2002)
Some novel triorganotin(IV) derivatives of β,δ-triketones.
Kumar A, et al.
Polyhedron, 2(9), 907-916 (1983)
Structure and magnetism of a binuclear 1, 3, 5-triketonate complex of cobalt (II), bis (1, 5-diphenyl-1, 3, 5-pentanetrionate) tetrapyridinedicobalt (II) tetrapyridine.
Kuszaj JM, et al.
Inorganic Chemistry, 12(6), 1297-1303 (1973)
Photochemical deposition of NiCoOx thin films from Ni/Co heteronuclear triketonate complexes.
Buono-Core GE, et al.
Materials Chemistry and Physics, 96(1), 98-102 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service