Skip to Content
Merck
All Photos(1)

Key Documents

42430

Sigma-Aldrich

N-Methyldioctylamine

≥96.0% (GC)

Synonym(s):

N,N-Dioctylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]2NCH3
CAS Number:
Molecular Weight:
255.48
Beilstein:
1750823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥96.0% (GC)

refractive index

n20/D 1.4424 (lit.)

bp

162-165 °C/15 mmHg (lit.)

mp

−30.1 °C (lit.)

density

0.793 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN(C)CCCCCCCC

InChI

1S/C17H37N/c1-4-6-8-10-12-14-16-18(3)17-15-13-11-9-7-5-2/h4-17H2,1-3H3

InChI key

YJLYANLCNIKXMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyldioctylamine (N,N,-dioctylmethylamine), a two branched C8 amine is an organic base.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

244.4 °F

Flash Point(C)

118 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Harding
The Biochemical journal, 117(5), 957-960 (1970-05-01)
Protein-bound glutathione was identified and measured in normal and cataractous human lenses. In a major group of cataracto us lenses the bound glutathione concentration was higher than normal. Study of normal lenses showed that their glutathione content is age-dependent, decreasing
G E Black et al.
Analytical chemistry, 66(23), 4171-4176 (1994-12-01)
Muramic acid is an amino sugar found in eubacterial cell walls and not elsewhere in nature. This study explored the use of electrospray tandem mass spectrometry (ESI MS/MS) in analysis of underivatized muramic acid in bacterial hydrolysates. Fungal hydrolysates were
Termite resistance of wood treated with copper (II) compounds derived from tri-and dialkylamine-boric acid complexes.
Chen GC, et al.
Forest products journal, 36(5), 18-20 (1986)
Development of reactive extraction systems for itaconic acid: a step towards in situ product recovery for itaconic acid fermentation.
Kaur G and Elst K.
Royal Society of Chemistry Advances, 4(85), 45029-45039 (2014)
Vanessa G Correia et al.
Macromolecular bioscience, 11(8), 1128-1137 (2011-06-16)
A method using supercritical CO(2) to obtain biocompatible 2-oxazoline-based oligomers quaternized with different amines is described. The synthesized oligo(2-oxazoline)s display partial carbamic-acid insertion at one end. The syntheses of quaternary oligo(2-bisoxazoline)s and linear oligoethylenimine hydrochlorides are reported. Oligo(2-methyl-2-oxazoline) and oligo(2-bisoxazoline)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service