Skip to Content
Merck
All Photos(1)

Key Documents

423831

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-L-leucine N′-methoxy-N′-methylamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH[NHCO2C(CH3)3]CON(OCH3)CH3
CAS Number:
Molecular Weight:
274.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]23/D −25.5°, c = 2 in methanol

refractive index

n20/D 1.454 (lit.)

bp

235 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CON(C)C(=O)[C@H](CC(C)C)NC(=O)OC(C)(C)C

InChI

1S/C13H26N2O4/c1-9(2)8-10(11(16)15(6)18-7)14-12(17)19-13(3,4)5/h9-10H,8H2,1-7H3,(H,14,17)/t10-/m0/s1

InChI key

IKRXSZUARJIXLZ-JTQLQIEISA-N

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service