Skip to Content
Merck
All Photos(2)

Key Documents

422746

Sigma-Aldrich

2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol

98%

Synonym(s):

2-(2-Hydroxy-3,5-di-tert -amylphenyl)-2H -benzotriazole, 2-(2H -Benzotriazol-2-yl)-4,6-bis(1,1-dimethylpropyl)phenol, 2-(3′,5′-Di-tert -amyl-2′-hydroxyphenyl)benzotriazole, 2-(3,5-Di-tert -amyl-2-hydroxyphenyl)-2H -benzotriazole, 2-[2-Hydroxy-3,5-bis(1,1-dimethylpropyl)phenyl]-2H -benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29N3O
CAS Number:
Molecular Weight:
351.49
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

80-83 °C (lit.)

SMILES string

CCC(C)(C)c1cc(c(O)c(c1)C(C)(C)CC)-n2nc3ccccc3n2

InChI

1S/C22H29N3O/c1-7-21(3,4)15-13-16(22(5,6)8-2)20(26)19(14-15)25-23-17-11-9-10-12-18(17)24-25/h9-14,26H,7-8H2,1-6H3

InChI key

ZMWRRFHBXARRRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol (UV-328) is an ultraviolet (UV) stabilizer with a phenolic group connected to the benzotriazole structure. These phenolic groups facilitate the absorption of UV light. UV-328 is mainly used in as a light stabilizer for coating and as an additive for industrial polymers.[1][2]

Application

UV-328 is a class of benzotriazole derivatives that can potentially be used as corrosion inhibitors in heat resistant fluids in aircraft, brake fluids in automobiles and for other industrial cooling systems.[3][4]

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah S D Lafleur et al.
Macromolecular rapid communications, 40(9), e1800811-e1800811 (2019-03-07)
Optical patterns are produced on the surface of drawn linear polyethylene containing 2-(2H-benzotriazol-2-yl)-4,6-ditertpentylphenol (BZT), a photothermal dye, by direct laser writing. The photothermal dye absorbs the UV light and dissipates heat in the polyethylene film. This heat locally results in
Stefan Weiss et al.
Analytical chemistry, 77(22), 7415-7420 (2005-11-16)
The first method for the determination of commonly used corrosion inhibitors in environmental water samples by liquid chromatography-electrospray ionization-tandem mass spectrometry is presented. Benzotriazole (BTri) and the two isomers of tolyltriazole (5- and 4-TTri) are separated in an isocratic run.
Determination of benzothiazole and benzotriazole derivates in tire and clothing textile samples by high performance liquid chromatography-electrospray ionization tandem mass spectrometry.
Avagyan R, et al.
Journal of Chromatography A, 1307(22), 119-125 (2013)
Corinna Fischer et al.
Toxicology in vitro : an international journal published in association with BIBRA, 68, 104932-104932 (2020-07-12)
The benzotriazole UV stabilizer (BUVS) 2-(5-chloro-benzotriazol-2-yl)-4,6-di-(tert-butyl)phenol (UV-327) is used in various plastic products to protect them against harmful UV radiation. Meanwhile, there are concerns about potential adverse health effects on humans, as residues of UV-327 and other BUVSs have already
Multiresidue analytical method for the determination of antimicrobials, preservatives, benzotriazole UV stabilizers, flame retardants and plasticizers in fish using ultra high performance liquid chromatography coupled with tandem mass spectrometry.
Kim J, et al.
Journal of Chromatography A, 1218(22), 3511-3520 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service