Skip to Content
Merck
All Photos(1)

Key Documents

420905

Supelco

QUILON® S chromium complex solution

5.5-6.0 wt. % Cr in isopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35Cl4Cr2O3
CAS Number:
Molecular Weight:
545.27
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

concentration

5.5-6.0 wt. % Cr in isopropanol

bp

~82 °C

mp

−35 °C

density

0.924 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCCCCCCC1=[O][Cr](Cl)(Cl)O[Cr](Cl)(Cl)O1

Legal Information

DuPont Product
QUILON is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service