Skip to Content
Merck
All Photos(1)

Key Documents

419990

Sigma-Aldrich

(1S)-(+)-Neomenthyl acetate

99%

Synonym(s):

(+)-(1S,2S,5R)-Neomenthyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
Beilstein:
2208504
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D +48°, c = 8 in benzene

bp

81 °C/3 mmHg (lit.)

mp

36-38 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@@H]1OC(C)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service