Skip to Content
Merck
All Photos(1)

Key Documents

404357

Sigma-Aldrich

2,4,5-Trifluorobenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2COCl
CAS Number:
Molecular Weight:
194.54
Beilstein:
3606259
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.498 (lit.)

density

1.52 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(F)c(cc1F)C(Cl)=O

InChI

1S/C7H2ClF3O/c8-7(12)3-1-5(10)6(11)2-4(3)9/h1-2H

InChI key

STBGCAUUOPNJBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,5-Trifluorobenzoyl chloride is an aryl fluorinated building block.

Application

2,4,5-Trifluorobenzoyl chloride may be used in the synthesis of several glycosylated derivatives of some fluoroquinolone antibiotics.[1] It may be employed in one-pot synthesis of ethyl 8,9-difluoro-6-oxo-6H-benzo[c]quinolizine-5-carboxylate.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis and biological activity of 8-fluoro-9-(4-methyl-1-piperazinyl)-6-oxo-6H-benzo[c]quinolizine-5-carboxylic acid.
Ziegler B, et al.
Journal of Medicinal Chemistry, 27(3), 587-589 (1990)
M E Jung et al.
Journal of medicinal chemistry, 42(19), 3899-3909 (1999-10-03)
We report herein the synthesis and biological testing of several glycosylated derivatives of some fluoroquinolone antibiotics. In particular, we have prepared several glycosylated derivatives of ciprofloxacin (2) in which the carbohydrate units are linked to the free secondary amine of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service