Skip to Content
Merck
All Photos(1)

Documents

398845

Sigma-Aldrich

Lead(IV) acetate

≥99.99% trace metals basis

Synonym(s):

Lead tetraacetate, Pb(acac)4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)4
CAS Number:
Molecular Weight:
443.38
Beilstein:
3595640
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99% trace metals basis

reaction suitability

core: lead
reagent type: catalyst

SMILES string

CC(=O)O[Pb](OC(C)=O)(OC(C)=O)OC(C)=O

InChI

1S/4C2H4O2.Pb/c4*1-2(3)4;/h4*1H3,(H,3,4);/q;;;;+4/p-4

InChI key

JEHCHYAKAXDFKV-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Smoothly induces the addition of difluorodiiodomethane to alkenes and alkynes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li, A-R. Chen, Q-Y.
Synthesis, 1481-1481 (1997)
T H Ma et al.
Mutation research, 270(1), 71-77 (1992-11-01)
Synergistic and antagonistic effects on genotoxicity of mixtures of four chemicals; i.e., lead tetraacetate (LTA), arsenic trioxide (ATO), dieldrin (DED), and tetrachloroethylene (TCE), were evaluated by the Tradescantia-micronucleus (Trad-MCN) assay. The chemicals were mixed in ratios of 1:1, 1:2 and
Glycol-cleavage oxidation.
Arthur S Perlin
Advances in carbohydrate chemistry and biochemistry, 60, 183-250 (2006-06-06)
Luis Moreno-Osorio et al.
Magnetic resonance in chemistry : MRC, 45(11), 993-996 (2007-09-19)
Two naturally occurring 11-nordrimanes were synthesized, and their (1)H and (13)C NMR spectra were unambiguously assigned in full for the first time.
Hidehito Urata et al.
Chemical & pharmaceutical bulletin, 56(11), 1611-1612 (2008-11-05)
The first chemical synthesis of two metabolites ((1R,2S,5R,6S)-6-(3,4-dihydroxyphenyl)-2-(3,4-methylenedioxyphenyl)-3,7-dioxabicyclo[3,3,0]octane (SC-1) and (1R,2S,5R,6S)-2,6-bis(3,4-dihydroxyphenyl)-3,7-dioxabicyclo[3,3,0]octane (SC-2)) of sesamin was achieved by a simple two-step approach from sesamin. The approach consists of acetoxylation of the methylenedioxy moiety(ies) with lead(IV) tetraacetate and acid hydrolysis of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service