Skip to Content
Merck
All Photos(1)

Key Documents

398233

Sigma-Aldrich

Magnesium bisulfite solution

30 wt. % in H2O

Synonym(s):

Magnesium hydrogen sulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(HSO3)2
CAS Number:
Molecular Weight:
186.45
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

concentration

30 wt. % in H2O

density

1.22 g/mL at 25 °C

SMILES string

[Mg++].OS([O-])=O.OS([O-])=O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hajdáková et al.
Folia microbiologica, 36(6), 578-581 (1991-01-01)
Since diluted magnesium bisulfite waste liquor from a pulp mill after beech wood processing exhibited an inhibitory effect on all the model organisms used in our previous work (Cernáková et al. 1991) and the results of Ames mutagenicity test were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service