Skip to Content
Merck
All Photos(1)

Documents

388238

Sigma-Aldrich

5-Acetyliminodibenzyl

99%

Synonym(s):

5-Acetyl-10,11-dihydro-5H-dibenz[b,f]azepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15NO
CAS Number:
Molecular Weight:
237.30
MDL number:
UNSPSC Code:
12352103

Assay

99%

mp

95-97 °C (lit.)

SMILES string

CC(=O)N1c2ccccc2CCc3ccccc13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service