Skip to Content
Merck
All Photos(1)

Key Documents

371777

Sigma-Aldrich

N-(4-Aminophenyl)-4-methylbenzenesulfonamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHC6H4NH2
CAS Number:
Molecular Weight:
262.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

183-185 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)Nc2ccc(N)cc2

InChI

1S/C13H14N2O2S/c1-10-2-8-13(9-3-10)18(16,17)15-12-6-4-11(14)5-7-12/h2-9,15H,14H2,1H3

InChI key

XKDFMOOUKPSLEM-UHFFFAOYSA-N

General description

N-(4-Aminophenyl)-4-methylbenzenesulfonamide is a sulfonamide derivative. It has been reported to crystallize in an asymmetric unit containing two independent molecules, having V-shaped conformations. Its crystals are orthorhombic having space group P212121.

Application

N-(4-Aminophenyl)-4-methylbenzenesulfonamide may be used in the preparation of following compounds:
  • 4-methyl-N-[4-[3-(2-naphthyl)imidazo[1,2-a]pyrazine-8-yl]aminophenyl]benzenesulfonamide
  • 4-methyl-N-[4-[2-(2-naphthyl)imidazo[1,2-a]pyrazine-8-yl]aminophenyl]benzenesulfonamide
  • 2- and 3-substituted 8-amino imidazo[1,2-a]pyrazines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeveria Rehman et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 10), o2709-o2709 (2011-11-09)
The title compound, C(13)H(14)N(2)O(2)S, crystallized with two independent mol-ecules in the asymmetric unit. They both have V-shaped conformations: the dihedral angles between their benzene rings are identical [45.86 (13)°] and their C-S-N-C torsion angles are similar [67.9 (3) and 70.2 (3)°]. In the
James R Sayer et al.
Bioorganic & medicinal chemistry, 22(22), 6459-6470 (2014-12-03)
A novel series of 8-amino imidazo[1,2-a]pyrazine derivatives has been developed as inhibitors of the VirB11 ATPase HP0525, a key component of the bacterial type IV secretion system. A flexible synthetic route to both 2- and 3-aryl substituted regioisomers has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service