Skip to Content
Merck
All Photos(1)

Key Documents

370606

Sigma-Aldrich

4-(Trifluoromethoxy)benzaldehyde

96%

Synonym(s):

α,α,α-Trifluoroanisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CHO
CAS Number:
Molecular Weight:
190.12
Beilstein:
1949135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

93 °C/27 mmHg (lit.)

density

1.331 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)Oc1ccc(C=O)cc1

InChI

1S/C8H5F3O2/c9-8(10,11)13-7-3-1-6(5-12)2-4-7/h1-5H

InChI key

XQNVDQZWOBPLQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Trifluoromethoxy)benzaldehyde (p-(trifluoromethoxy)benzaldehyde) is an aromatic aldehyde. It participates in the synthesis of 4,5-dihydro-4-(4-trifluoromethoxyphenyl)pyrrolo[1,2-a]quinoxaline.[1]

Application

4-(Trifluoromethoxy)benzaldehyde is suitable for use in the synthesis of (E)-4-(trifluoromethoxy)benzaldehyde thiosemicarbazone.[2] It may be used in the synthesis of:
  • new azo dyes containing of 5(4H)-oxazolone ring[3]
  • 2-[4-(trifluoromethoxy)phenyl]-1H-benzimidazole[4]
  • (E)-2-[4-(trifluoromethoxy)benzylidene]indan-1-one.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikhath Fathima et al.
Acta crystallographica. Section E, Structure reports online, 69(Pt 2), o264-o264 (2013-02-21)
In the title compound, C(14)H(9)F(3)N(2)O, the best planes of the benzimidazole group and benzene ring form a dihedral angle of 26.68 (3)°. In the crystal, N-H⋯N hydrogen bonds link the mol-ecules into infinite chains parallel to the c axis. Stacking inter-actions
Andreas J Kesel
European journal of medicinal chemistry, 46(5), 1656-1664 (2011-03-08)
Aromatic aldehyde-derived thiosemicarbazones 4-6, the S-substituted modified thiosemicarbazones 7/8, and a vitamin A-derived (retinoid) thiosemicarbazone derivative 12 were investigated as inhibitors of human hepatitis C virus (HCV) subgenomic RNA replicon Huh7 ET (luc-ubi-neo/ET) replication. Compounds 4-6 and 12 were found
Mohamed Ashraf Ali et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 8), o2147-o2147 (2011-11-18)
In the title compound, C(17)H(11)F(3)O(2), the dihydro-indene ring is approximately planar with a maximum deviation of 0.024 (2) Å and makes a dihedral angle of 3.17 (8) Å with the adjacent benzene ring. In the crystal, mol-ecules are inter-connected by C-H⋯O inter-actions, forming an
A Versatile Synthesis of 4, 5-Dihydropyrrolo [1, 2-a] quinoxalines.
Abonia R, et al.
Journal of Heterocyclic Chemistry, 38(3), 671-674 (2001)
Hooshang Hamidian et al.
Bioorganic & medicinal chemistry, 21(7), 2088-2092 (2013-02-16)
Six new azo dyes containing of 5(4H)-oxazolone ring were prepared by diazotization of 4-aminohippuric acid and coupling with N,N-dimethylaniline, 1-naphthol and 2-naphthol and condensation with 4-fluoro benzaldehyde or 4-trifluoromethoxy benzaldehyde. The new compounds were fully characterized by spectroscopic techniques. All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service