Skip to Content
Merck
All Photos(2)

Documents

370479

Sigma-Aldrich

Diethyl (2-oxopropyl)phosphonate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
194.17
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.433 (lit.)

bp

126 °C/9 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CC(C)=O)OCC

InChI

1S/C7H15O4P/c1-4-10-12(9,11-5-2)6-7(3)8/h4-6H2,1-3H3

InChI key

RSAFKRSMGOSHRK-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service