All Photos(1)
About This Item
Linear Formula:
F2C6H2(OCH3)2
CAS Number:
Molecular Weight:
174.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
90%
form
solid
mp
41-43 °C (lit.)
density
1.226 g/mL at 25 °C (lit.)
SMILES string
COc1cc(F)c(F)cc1OC
InChI
1S/C8H8F2O2/c1-11-7-3-5(9)6(10)4-8(7)12-2/h3-4H,1-2H3
InChI key
WWAOVLXLTJXDGS-UHFFFAOYSA-N
General description
1,2-Difluoro-4,5-dimethoxybenzene is an aryl fluorinated building block.
Application
1,2-Difluoro-4,5-dimethoxybenzene may be used in the synthesis of molecular crystals of [Li{N(SO2CF3)2}{C6H4(OCH3)2}2] and [Li{N(SO2CF3)2}{C6F2H2(OCH3)2}2], having solid-state lithium ion conductivity.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
197.6 °F - closed cup
Flash Point(C)
92 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Makoto Moriya et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(40), 13554-13560 (2013-08-14)
The molecular crystals [Li{N(SO2CF3)2}{C6H4(OCH3)2}2] and [Li{N(SO2CF3)2}{C6F2H2(OCH3)2}2] with solid-state lithium ion conductivity have been synthesized by the addition of two equivalents of 1,2-dimethoxybenzene or 1,2-difluoro-4,5-dimethoxybenzene to Li{N(SO2CF3)2}, respectively. Single-crystal X-ray diffraction analysis revealed the formation of ionic conduction paths with an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service