Skip to Content
Merck
All Photos(1)

Key Documents

367834

Sigma-Aldrich

1,4-Diaminoanthraquinone

technical grade, ≥88.0%

Synonym(s):

1,4-Diamino-9,10-anthracenedione, 1,4-Diamino-9,10-anthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O2
CAS Number:
Molecular Weight:
238.24
Beilstein:
2216556
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

≥88.0%

mp

265-269 °C (lit.)

SMILES string

Nc1ccc(N)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C14H10N2O2/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6H,15-16H2

InChI key

FBMQNRKSAWNXBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

644.0 °F - closed cup

Flash Point(C)

340 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan P May et al.
Chemical communications (Cambridge, England), (8)(8), 970-971 (2003-05-15)
A novel, long-wavelength, non-fluorescent quencher (LQ), based on 1,4-diaminoanthraquinone, has been incorporated at the 3' and 5'-termini of oligonucleotides. The quencher has been used in Molecular beacons, efficiently quenching the long wavelength fluorophore, Cy5.
Abhishek Mandal et al.
Inorganic chemistry, 53(12), 6082-6093 (2014-06-05)
The compounds [(acac)2Ru(III)(μ-H2L(2-))Ru(III)(acac)2] (rac, 1, and meso, 1') and [(bpy)2Ru(II)(μ-H2L(•-))Ru(II)(bpy)2](ClO4)3 (meso, [2](ClO4)3) have been structurally, magnetically, spectroelectrochemically, and computationally characterized (acac(-) = acetylacetonate, bpy = 2,2'-bipyridine, and H4L = 1,4-diamino-9,10-anthraquinone). The N,O;N',O'-coordinated μ-H2L(n-) forms two β-ketiminato-type chelate rings, and 1
Mohammad Saeid Hosseini et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 25(6), 807-812 (2009-06-18)
A highly sensitive and selective method has been proposed for speciation determination of trace amounts of Cr(III) and Cr(VI) in water samples using 1,4-diaminoanthraquinone (1,4-DAAQ). The method is based on mixing an organic phase containing 1,4-DAAQ with an acidic solution
Vsevolod Katritch et al.
Journal of medicinal chemistry, 53(4), 1799-1809 (2010-01-26)
The recent progress in crystallography of G-protein coupled receptors opens an unprecedented venue for structure-based GPCR drug discovery. To test efficiency of the structure-based approach, we performed molecular docking and virtual ligand screening (VLS) of more than 4 million commercially
The mutagenic effect in bacteriophage T4D of a hair dye, 1,4 diaminoanthraquinone, and of two solvents, dimethylsulfoxide and ethanol.
I Kvelland
Hereditas, 99(2), 209-213 (1983-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service