Skip to Content
Merck
All Photos(1)

Key Documents

363731

Sigma-Aldrich

3′-Chloroacetanilide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4Cl
CAS Number:
Molecular Weight:
169.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

mp

79-81 °C (lit.)

SMILES string

CC(=O)Nc1cccc(Cl)c1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Carrera et al.
Archives of environmental contamination and toxicology, 35(1), 89-96 (1998-06-17)
The changes in metabolism and cytotoxicity of chlorpropham (CIPC) and its major metabolites, 4-hydroxychlorpropham (4-OH CIPC), 3-chloroaniline, and 3-chloroacetanilide were investigated in isolated rat hepatocyte suspensions after a partial inhibition of sulphation and glucuronidation and the two reactions combined in
X Q Wang et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(1), 59-69 (1994-01-01)
1. Guppies (Poecilia reticulata) were exposed for 96 h to 4-chloroaniline (4CA), 3-chloroaniline (3CA) and 2-chloroaniline (2CA) in a static exposure system. 4-Chloroacetanilide (N4CA), 3-chloroacetanilide (N3CA), and 2-chloroacetanilide (N2CA) were identified in test water as the respective biotransformation products. 2.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service