Skip to Content
Merck
All Photos(1)

Key Documents

344125

Sigma-Aldrich

(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine

97%

Synonym(s):

(R)-Ugi′s amine, (R)-(+)-[1-(Dimethylamino)ethyl]ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19FeN
CAS Number:
Molecular Weight:
257.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D +13°, c = 1 in ethanol (5 h)

refractive index

n20/D 1.589 (lit.)

bp

120-121 °C/0.7 mmHg (lit.)

density

1.222 g/mL at 20 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][CH]2)N(C)C

InChI

1S/C9H14N.C5H5.Fe/c1-8(10(2)3)9-6-4-5-7-9;1-2-4-5-3-1;/h4-8H,1-3H3;1-5H;/t8-;;/m1../s1

InChI key

ISOLNZQTVMCEOI-YCBDHFTFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine may be used as a starting material to synthesize:
  • chiral 1,2-disubstituted ferrocenyl amino alcohols[1]
  • ferrocene-based P-chiral phosphine ligands[2]
  • optically active α-ferrocenyl tertiary amines[3]
Ligand used in the preparation of chiral allylic alcohols,[4] axially chiral allenecarboxylic esters,[5] and rhodium(I)-catalyzed asymmetric hydrosilylation of ketones.[6]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis. VIII. Absolute configuration of a 1, 2-disubstituted ferrocene derivative with planar and central elements of chirality and the mechanism of the optically active. alpha.-ferrocenyl tertiary amines.
Battelle LF, et al.
Journal of the American Chemical Society, 95(2), 482-486 (1973)
Chiba, T. et al.
Tetrahedron Letters, 36, 1519-1519 (1995)
Chiral auxiliaries for asymmetric synthesis: enantioselective addition of dialkylzincs to aldehydes catalyzed by chiral 1, 2-disubstituted ferrocenyl amino alcohols.
Watanabe M, et al.
The Journal of Organic Chemistry, 56(6), 2218-2224 (1991)
Nishibayashi, Y. et al.
Tetrahedron Letters, 35, 3115-3115 (1994)
Nishibayashi, Y.et al.
Journal of the Chemical Society. Chemical Communications, 1375-1375 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service