Skip to Content
Merck
All Photos(1)

Key Documents

343811

Sigma-Aldrich

Ethylene glycol-(OD)2

98 atom % D, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DOCH2CH2OD
CAS Number:
Molecular Weight:
64.08
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
technique(s):
NMR: suitable
bp:
196-198 °C (lit.)
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.429 (lit.)

bp

196-198 °C (lit.)

mp

-13 °C (lit.)

density

1.148 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]OCCO[2H]

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2/i3D,4D

InChI key

LYCAIKOWRPUZTN-NMQOAUCRSA-N

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service