Skip to Content
Merck
All Photos(1)

Key Documents

328200

Sigma-Aldrich

2,4,5-Trifluorobenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2CN
CAS Number:
Molecular Weight:
157.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.4736 (lit.)

density

1.373 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

Fc1cc(F)c(cc1F)C#N

InChI

1S/C7H2F3N/c8-5-2-7(10)6(9)1-4(5)3-11/h1-2H

InChI key

DLKNOGQOOZFICZ-UHFFFAOYSA-N

General description

Raman and FTIR spectra of 2,4,5- Trifluorobenzonitrile was studied.[1] Highly regioselective substitution reaction of 2,4,5-trifluorobenzonitrile was reported.[2]

Application

2,4,5-Trifluorobenzonitrile was used in preparation of novel linker in solid phase synthesis, 2-(2-fluoro-4-hydroxymethyl-5-methoxy-phenoxy)acetic acid.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective nucleophilic substitutions of fluorobenzene derivatives
Wells KM, et al.
Tetrahedron Letters, 37(36), 6439-6442 (1996)
V Mukherjee et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(4), 1571-1580 (2008-08-12)
Raman and FTIR spectra of 2,4,5- and 3,4,5-tri-fluoro-benzonitriles have been recorded in the regions 50-4000 cm(-1) and 400-4000 cm(-1), respectively. Measurement of depolarization ratios for the Raman lines has also been made. Optimized geometrical parameters, charge distributions and vibrational wavenumbers
Fredrik K Wallner et al.
Organic & biomolecular chemistry, 5(15), 2464-2471 (2007-07-20)
Gel-phase (19)F NMR spectroscopy is a useful monitoring technique for solid-phase organic chemistry due to the high information content it delivers and swift acquisition times, using standard NMR spectrometers. This paper describes the synthesis of the novel linker 2-(2-fluoro-4-hydroxymethyl-5-methoxy-phenoxy)acetic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service