Recommended Products
Assay
98%
mp
62-64 °C (lit.)
SMILES string
COc1ccc(F)cc1[N+]([O-])=O
InChI
1S/C7H6FNO3/c1-12-7-3-2-5(8)4-6(7)9(10)11/h2-4H,1H3
InChI key
FWLPYISRFBKEKV-UHFFFAOYSA-N
General description
4-Fluoro-2-nitroanisole is formed during the photolysis of the charge transfer complex of tetranitromethane and 4-fluoroanisole in dichloromethane at -20°C.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Photochemical nitration by tetranitromethane. Part 36. Adduct formation in the photochemical reactions of 4-fluoroanisole and 4-fluoro-3-methylanisole.
Journal of the Chemical Society. Perkin Transactions 1, 2(9), 1877-1887 (1996)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service