Skip to Content
Merck
All Photos(1)

Key Documents

316296

Sigma-Aldrich

4-Bromo-2-fluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)OH
CAS Number:
Molecular Weight:
191.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 316296, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

liquid

refractive index

n20/D 1.566 (lit.)

bp

79 °C/7 mmHg (lit.)

density

1.744 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Oc1ccc(Br)cc1F

InChI

1S/C6H4BrFO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

RYVOZMPTISNBDB-UHFFFAOYSA-N

Application

4-Bromo-2-fluorophenol can be used as a starting material for the synthesis of:
  • 4-Bromo-2-fluoro-6-iodoanisole via iodination and methylation.[1]
  • 2-Phenylpyran-4-ones for active cyclooxygenase-2 inhibitor evaluation.[2]
  • 1,4-Disubstituted 3-cyano-2-pyridones.[3]
  • Dicationic imidazolium-based compounds.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and Reliable Iodination and O-Methylation of Fluorinated Phenols
Francke R, et al.
European Journal of Organic Chemistry, 2010(12), 2357-2362 (2010)
Discovery of 1, 4-disubstituted 3-cyano-2-pyridones: a new class of positive allosteric modulators of the metabotropic glutamate 2 receptor
Cid JM, et al.
Journal of Medicinal Chemistry, 55(5), 2388-2405 (2012)
Dicationic imidazolium-based ionic liquids and ionic liquid crystals with variously positioned fluoro substituents
Li X, et al.
Journal of Materials Chemistry, 19(43), 8232-8238 (2009)
Synthesis and biological evaluation of 2-phenylpyran-4-ones: a new class of orally active cyclooxygenase-2 inhibitors
Caturla F, et al.
Journal of Medicinal Chemistry, 47(15), 3874-3886 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service