About This Item
Recommended Products
Assay
98%
SMILES string
O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])=O
InChI
1S/2Na.H2O3Se.5H2O/c;;1-4(2)3;;;;;/h;;(H2,1,2,3);5*1H2/q2*+1;;;;;;/p-2
InChI key
TUANAMBRHOLYTH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service