Skip to Content
Merck
All Photos(2)

Key Documents

303895

Sigma-Aldrich

4-Chromanol

≥99%

Synonym(s):

3,4-Dihydro-2H-1-benzopyran-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

form

solid

mp

42-44 °C (lit.)

functional group

hydroxyl

SMILES string

OC1CCOc2ccccc12

InChI

1S/C9H10O2/c10-8-5-6-11-9-4-2-1-3-7(8)9/h1-4,8,10H,5-6H2

InChI key

MGSHXMOLUWTMGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chromanol was used in preparation of 4-α-L-rhamnopyranosyloxychromanol, glycosyl flavonoid, by glycosylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoliang Yang et al.
Organic & biomolecular chemistry, 4(12), 2483-2491 (2006-06-10)
Astilbin, a flavonoid isolated from different plants, shows diverse biological activities. This paper reports the synthesis and immunosuppressive activity of seven analogues of astilbin, which may shed light on the structure-activity relationship of the compounds. The following glycosyl flavonoids, 6-alpha-L-rhamnopyranosyloxyflavanone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service