Skip to Content
Merck
All Photos(1)

Key Documents

300799

Sigma-Aldrich

3-Methoxy-N,N-dimethylbenzylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2N(CH3)2
CAS Number:
Molecular Weight:
165.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

refractive index

n20/D 1.514 (lit.)

bp

105 °C/13 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

COc1cccc(CN(C)C)c1

InChI

1S/C10H15NO/c1-11(2)8-9-5-4-6-10(7-9)12-3/h4-7H,8H2,1-3H3

InChI key

IENUHHADGQLQLU-UHFFFAOYSA-N

Application

3-Methoxy-N,N-dimethylbenzylamine was used in the preparation of ethyl 6-chloromethyl-2-methoxybenzoate[1] and 2-lithio-N,N-dimethylbenzylamine (five-membered-ring amine chelated aryllithium reagent).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junia M Pereira et al.
Bioorganic & medicinal chemistry, 16(19), 8889-8895 (2008-09-16)
Chagas' disease, a parasitic infection caused by the flagellate protozoan Trypanosoma cruzi, is a major public health problem affecting millions of individuals in Latin America. On the basis of the essential role in the life cycle of T. cruzi, the
H J Reich et al.
Journal of the American Chemical Society, 123(33), 8067-8079 (2001-08-17)
Multinuclear NMR studies of five-membered-ring amine chelated aryllithium reagents 2-lithio-N,N-dimethylbenzylamine (1), the diethylamine and diisopropylamino analogues (2, 3), and the o-methoxy analogue (4), isotopically enriched in (6)Li and (15)N, have provided a detailed picture of the solution structures in ethereal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service