Skip to Content
Merck
All Photos(1)

Key Documents

295728

Sulfur tetrafluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SF4
CAS Number:
Molecular Weight:
108.06
EC Number:
MDL number:
Pricing and availability is not currently available.

vapor pressure

140 psi ( 21 °C)

bp

−40.4 °C (lit.)

mp

−121.5-−120.5 °C (lit.)

SMILES string

FS(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Application

Has found widespread use in the synthesis of fluoroalkyl groups[1] particularly from carbonyl functionality. Recent applications include fluorination of methyl groups[2] and oxo esters.[3]

Recommended products

The use of stainless steel control valve Z146951 or either one of stainless steel regulators Z148504 and Z148512 is recommended

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Fluorine Chemistry, 75, 115-115 (1995)
Journal of Fluorine Chemistry, 74, 259-259 (1995)
Aldrichimica Acta, 21, 3-3 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service