Skip to Content
Merck
All Photos(1)

Documents

284904

Sigma-Aldrich

Piperazine dihydrochloride hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10N2 · 2HCl · xH2O
CAS Number:
Molecular Weight:
159.06 (anhydrous basis)
Beilstein:
3551215
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

SMILES string

Cl[H].Cl[H].[H]O[H].C1CNCCN1

InChI

1S/C4H10N2.2ClH.H2O/c1-2-6-4-3-5-1;;;/h5-6H,1-4H2;2*1H;1H2

InChI key

ZQVBNUIFDLXAFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service