Skip to Content
Merck
All Photos(1)

Key Documents

281425

Sigma-Aldrich

6,7-Dichloro-1,4-dihydroxyanthraquinone

97%

Synonym(s):

6,7-Dichloroquinizarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6Cl2O4
CAS Number:
Molecular Weight:
309.10
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

97%

mp

290 °C (dec.) (lit.)

SMILES string

Oc1ccc(O)c2C(=O)c3cc(Cl)c(Cl)cc3C(=O)c12


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service