Skip to Content
Merck
All Photos(1)

Key Documents

278912

Sigma-Aldrich

3-(Trimethylsilyl)-2-oxazolidinone

98%

Synonym(s):

2-Oxo-3-(trimethylsilyl)-1,3-oxazolidine, TMSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2Si
CAS Number:
Molecular Weight:
159.26
Beilstein:
1210407
MDL number:
Pricing and availability is not currently available.

Assay

98%

density

1.046 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)N1CCOC1=O

InChI

1S/C6H13NO2Si/c1-10(2,3)7-4-5-9-6(7)8/h4-5H2,1-3H3

InChI key

AUKCYOUETBBMFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silylation reagent for hydroxyls.[1][2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canadian Journal of Chemistry, 62, 336-336 (1984)
Tec. Lab., 15, 506-506 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service