Skip to Content
Merck
All Photos(1)

Documents

276189

Sigma-Aldrich

Trimethylsilyl trimethylsiloxyacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOCH2CO2Si(CH3)3
CAS Number:
Molecular Weight:
220.41
Beilstein:
2045011
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.411 (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OCC(=O)O[Si](C)(C)C

InChI

1S/C8H20O3Si2/c1-12(2,3)10-7-8(9)11-13(4,5)6/h7H2,1-6H3

InChI key

MAEQOWMWOCEXKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylsilyl trimethylsiloxyacetate has been used in the synthesis of zirconium(IV) carboxylato complexes [LOEtZr(OCOCH2OH)(O2CCH2O)]2.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structures of zirconium (IV) hydrogensulfato and carboxylato complexes with Klaui's oxygen tripodal ligand.
Yi X-Y, et al.
Journal of Organometallic Chemistry, 694(26), 4256-4260 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service