Skip to Content
Merck
All Photos(1)

Key Documents

273244

Sigma-Aldrich

3′-Methylacetanilide

98%

Synonym(s):

m-Acetotoluidide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4CH3
CAS Number:
Molecular Weight:
149.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

16 mmHg ( 163 °C)

Assay

98%

bp

303 °C (lit.)

mp

65-67 °C (lit.)

solubility

alcohol: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
water: slightly soluble(lit.)

SMILES string

CC(=O)Nc1cccc(C)c1

InChI

1S/C9H11NO/c1-7-4-3-5-9(6-7)10-8(2)11/h3-6H,1-2H3,(H,10,11)

InChI key

ALMHSXDYCFOZQD-UHFFFAOYSA-N

General description

The IR spectra and the structural data of 3′-methylacetanilide was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wioleta Rmiszek-Lindert et al.
Acta crystallographica. Section C, Crystal structure communications, 64(Pt 11), o599-o603 (2008-11-08)
Molecules of the title compounds, 3'-methylacetanilide [or N-(m-tolyl)acetamide], C(9)H(11)NO, (I), and N-benzylthioacetamide, C(9)H(11)NS, (II), are connected by a framework of intermolecular N-H...O and N-H...S hydrogen bonds, respectively, forming chains with the graph-set description C(4), which run along the b axis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service