Skip to Content
Merck
All Photos(1)

Key Documents

262331

Sigma-Aldrich

Pentyltrichlorosilane,mixture of isomers

97%

Synonym(s):

Amyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

97%


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service