Recommended Products
vapor pressure
20 mmHg ( 100 °C)
Assay
95%
refractive index
n20/D 1.429 (lit.)
bp
94 °C/15 mmHg (lit.)
density
0.84 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCC1CO1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service