Skip to Content
Merck
All Photos(1)

Key Documents

26033

Sigma-Aldrich

2-Chlorophenyl 2,2,2-trichloroethyl chlorophosphate

purum, ≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6Cl5O3P
CAS Number:
Molecular Weight:
358.37
Beilstein:
2005867
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (AT)

refractive index

n20/D 1.535

density

1.574 g/mL at 20 °C (lit.)

SMILES string

Clc1ccccc1OP(Cl)(=O)OCC(Cl)(Cl)Cl

Other Notes

Phosphorylating agent for preparing protected nucleotides

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Wolter et al.
Tetrahedron Letters, 24, 873-873 (1983)
J.H. van Boom et al.
Tetrahedron, 34, 1999-1999 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service