Skip to Content
Merck
All Photos(1)

Key Documents

250392

Sigma-Aldrich

3-Hexyne-2,5-diol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)C≡CCH(OH)CH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
1720667
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.473 (lit.)

bp

121 °C/15 mmHg (lit.)

mp

42 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

SMILES string

CC(O)C#CC(C)O

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h5-8H,1-2H3

InChI key

KDOWHHULNTXTNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

253.4 °F - closed cup

Flash Point(C)

123 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service