Skip to Content
Merck
All Photos(1)

Key Documents

241792

Sigma-Aldrich

3-Nitro-1H-1,2,4-triazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2N4O2
CAS Number:
Molecular Weight:
114.06
Beilstein:
607167
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

210 °C (dec.) (lit.)

SMILES string

[O-][N+](=O)c1nc[nH]n1

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitro-1H-1,2,4-triazole was used to detect trace amounts of mercuric ion in aqueous solution[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojun Chen et al.
The Analyst, 136(8), 1690-1696 (2011-03-04)
A simple colorimetric assay with high sensitivity, excellent selectivity and a tunable dynamic range is reported for detecting trace amounts of mercuric ion in aqueous solution based on the coordination of Hg(2+) to the gold nanoparticle (AuNP)-associated 3-nitro-1H-1,2,4-triazole (NTA). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service