Skip to Content
Merck
All Photos(1)

Key Documents

237477

Sigma-Aldrich

(3,4-Dimethoxyphenyl)acetyl chloride

98%

Synonym(s):

Homoveratryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2COCl
CAS Number:
Molecular Weight:
214.65
Beilstein:
396343
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.5489 (lit.)

bp

118-119 °C/0.2 mmHg (lit.)

mp

40-43 °C (lit.)

density

1.245 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CC(Cl)=O)cc1OC

InChI

1S/C10H11ClO3/c1-13-8-4-3-7(6-10(11)12)5-9(8)14-2/h3-5H,6H2,1-2H3

InChI key

QBJIMTPENIGDOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(3,4-Dimethoxyphenyl)acetyl chloride (Homoveratryl chloride) has been used in the preparation of:
  • homo veratryl nitrile (HVN)[1]
  • isomeric naphthalene sulphonyl compounds, 1- and 2-[{2-(3,4-dimethoxyphenyl)ethyl}methylamino] sulphonyl naphthalenes[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and antimicrobial activity studies of 1-& 2-[{2-(3, 4-dimethoxyphenyl) ethyl}-methylamino] sulphonyl naphthalenes.
Dayalan A, et al.
Asian Journal of Chemistry, 20(2), 1411-1419 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service