Skip to Content
Merck
All Photos(1)

Documents

226793

Sigma-Aldrich

cis-Decahydronaphthalene-d18

98 atom % D

Synonym(s):

cis-Decalin-d18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D18
CAS Number:
Molecular Weight:
156.36
MDL number:
UNSPSC Code:
12352200

isotopic purity

98 atom % D

mol wt

156.40

refractive index

n20/D 1.4761 (lit.)

bp

193 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

mass shift

M+18

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])[C@@]2([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[C@@]2([2H])C1([2H])[2H]


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service