Skip to Content
Merck
All Photos(1)

Key Documents

226106

Sigma-Aldrich

1,3-Bis(trimethylsilyl)urea

95%

Synonym(s):

N,N′-Bis(trimethylsilyl)urea, BSU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiNHCONHSi(CH3)3
CAS Number:
Molecular Weight:
204.42
Beilstein:
1937452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

219-221 °C (lit.)

solubility

pyridine: 100 mg/mL, clear to slightly hazy, colorless to light yellow

functional group

amine

SMILES string

C[Si](C)(C)NC(=O)N[Si](C)(C)C

InChI

1S/C7H20N2OSi2/c1-11(2,3)8-7(10)9-12(4,5)6/h1-6H3,(H2,8,9,10)

InChI key

MASDFXZJIDNRTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient method for the t-butyldimethylsilylation of alcohols with N,O-bis (t-butyldimethylsilyl) acetamide.
Johnson DA and Taubner LM.
Tetrahedron Letters, 37(5), 605-608 (1996)
Hung-Cuong Trinh et al.
Bio Systems, 137, 64-72 (2015-08-16)
Efficiently identifying functionally important genes in order to understand the minimal requirements of normal cellular development is challenging. To this end, a variety of structural measures have been proposed and their effectiveness has been investigated in recent literature; however, few

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service