Skip to Content
Merck
All Photos(1)

Key Documents

219118

Sigma-Aldrich

D-Asparagine monohydrate

99%

Synonym(s):

(R)-(-)-2-Aminosuccinamic acid, (R)-2-Aminosuccinic acid 4-amide, (D)-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COCH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723526
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D −27°, c = 1 in 1 M HCl

mp

275 °C (dec.) (lit.)

SMILES string

NC(C[C@@H](N)C(O)=O)=O.[H]O[H]

InChI

1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m1./s1

InChI key

RBMGJIZCEWRQES-HSHFZTNMSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service