Skip to Content
Merck
All Photos(2)

Key Documents

204242

Sigma-Aldrich

Bis(triphenylphosphine)rhodium(I) carbonyl chloride

99.9%

Synonym(s):

RhCl(CO)(PPh3)2, Chlorocarbonylbis(triphenylphosphine)rhodium(I), Rhodiumbis(triphenylphosphine) carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2RhCl(CO)
CAS Number:
Molecular Weight:
690.94
Beilstein:
4618896
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

99.9%

reaction suitability

core: rhodium
reagent type: catalyst

mp

224-227 °C (dec.) (lit.)

SMILES string

Cl[Rh].[C-]#[O+].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.CO.ClH.Rh/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2;;/h2*1-15H;;1H;/q;;;;+1/p-1

InChI key

SGULOPRLUCGLSH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used for cyclometallation.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiang, X.; Shengming, M. J.
Journal of the American Chemical Society, 129 (2007)
Xuefeng Jiang et al.
Journal of the American Chemical Society, 129(37), 11600-11607 (2007-08-30)
Cyclometallation of two unsaturated carbon-carbon bonds usually requires the application of low-valent metal catalysts, which could cleave the propargylic ester linkage. Thus, it is desirable to identify a catalyst which could undergo cyclometallation without cleaving the propargylic ester linkage. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service